Learn More
Enterprise Technology Solutions Midlab Inc Cleaner/Degreaser,All-Purpose,1 Gallon,4/CT,WE
SDP
Catalog No. 5000790005
Encompass_Preferred
All-Purpose Butyl-Based Cleaner and Degreaser easily removes particulates and oily soils from virtually any washable surface. Low-foaming concentrate is ideal when rinsing is not feasible so it works great in automatic floor-scrubbing machines. It is safe to use on soft metals so you can use it to clean coils. No rinsing is necessary.
Catalog No. 50-007-90005 Supplier Enterprise Technology Solutions Supplier No. MLB06100041
All-Purpose Butyl-Based Cleaner and Degreaser easily removes particulates and oily soils from virtually any washable surface. Low-foaming concentrate is ideal when rinsing is not feasible so it works great in automatic floor-scrubbing machines. It is safe to use on soft metals so you can use it to clean coils. No rinsing is necessary.