Learn More
Enterprise Technology Solutions Midlab Inc Cleaner/Degreaser, Lemon, RTU, 1 Quart, 12/CT, Yellow
SDP
Catalog No. 5000790058
Encompass_Preferred
Bulldozer Cleaner/Degreaser RTU contains a super-strong formula that removes grease, dirt, mildew stains, soap scum, wax and other soils from most water-washable surfaces. Spray and wipe fiberglass, linoleum, formica, vinyl, carpets, porcelain and chrome. This multipurpose, ready-to-use cleaner/degreaser also works on tubs, bathrooms, kitchens, engines, white sidewall tires, filters, floor mats, vinyl siding, tile, stoves, hoods, ovens, lawn mowers and golf carts. For vinyl tops and seats, agitate with sponge or cloth only and avoid dwell time on painted surfaces. Use as a spot remover on some surfaces. Do not use on glass or aluminum. Versatile cleaner has a lemon fragrance.
Catalog No. 50-007-90058 Supplier Enterprise Technology Solutions Supplier No. MLB05100012
Bulldozer Cleaner/Degreaser RTU contains a super-strong formula that removes grease, dirt, mildew stains, soap scum, wax and other soils from most water-washable surfaces. Spray and wipe fiberglass, linoleum, formica, vinyl, carpets, porcelain and chrome. This multipurpose, ready-to-use cleaner/degreaser also works on tubs, bathrooms, kitchens, engines, white sidewall tires, filters, floor mats, vinyl siding, tile, stoves, hoods, ovens, lawn mowers and golf carts. For vinyl tops and seats, agitate with sponge or cloth only and avoid dwell time on painted surfaces. Use as a spot remover on some surfaces. Do not use on glass or aluminum. Versatile cleaner has a lemon fragrance.