Learn More
Enterprise Technology Solutions Midlab Inc Cleaner,f/Glass,Concentrate,64oz,4/CT,BE
SDP
Catalog No. 5000790661
Encompass_Preferred
Sparkle Glass Cleaner delivers fast, easy cleaning and excellent streak-free results. It quickly removes dirt, dust, fingerprints and other light soils. Use this versatile cleaner on windows, metal, plastic, fiberglass and other hard surfaces. It dries quickly with a minimum of wiping. Eco-friendly choice contains no ammonia or fragrances. Dilution ratio is 1 oz. per gallon.
Catalog No. 50-007-90661 Supplier Enterprise Technology Solutions Supplier No. MLB95330065T
Sparkle Glass Cleaner delivers fast, easy cleaning and excellent streak-free results. It quickly removes dirt, dust, fingerprints and other light soils. Use this versatile cleaner on windows, metal, plastic, fiberglass and other hard surfaces. It dries quickly with a minimum of wiping. Eco-friendly choice contains no ammonia or fragrances. Dilution ratio is 1 oz. per gallon.