Learn More
Enterprise Technology Solutions Midlab Inc Floor Cleaner,Digester,2.6 oz Packets,30/Carton,WE
SDP
Catalog No. 5000790604
Encompass_Preferred
Fresh Traction Wash is a 2-in-1 cleaner and fresh digester, delivering odor-free, soil-free floors and grout. Kitchen floors require special attention due to the types of soils they're exposed to, including animal fats, vegetable oils and normal, tracked in dirt. Kitchen floors, especially those with mortar grout, become slippery and hard to clean. Heavy-duty cleaner with a neutral pH features enzymes that clean the floor and eliminate unpleasant odors, improving slip-resistance for worry-free floors.
Catalog No. 50-007-90604 Supplier Enterprise Technology Solutions Supplier No. MLB26040069
Fresh Traction Wash is a 2-in-1 cleaner and fresh digester, delivering odor-free, soil-free floors and grout. Kitchen floors require special attention due to the types of soils they're exposed to, including animal fats, vegetable oils and normal, tracked in dirt. Kitchen floors, especially those with mortar grout, become slippery and hard to clean. Heavy-duty cleaner with a neutral pH features enzymes that clean the floor and eliminate unpleasant odors, improving slip-resistance for worry-free floors.