Learn More
Enterprise Technology Solutions Midlab Inc LVT Daily Floor Cleaner Plus, Concentrate, 1 Gallon, 4/CT, Pale Red
SDP
Catalog No. 5000790061
Encompass_Preferred
LVT Daily Floor Cleaner Plus is a daily cleaner for low maintenance. It both cleans and continually repairs abrasions without building gloss or leaving a film or tacky residue. Apply solution with a mop or autoscrubber. Dilute using 2 oz. with 1 gallon of water. Use in schools, offices, supermarkets, hospitals and medical centers.
Catalog No. 50-007-90061 Supplier Enterprise Technology Solutions Supplier No. MLB17000065T
LVT Daily Floor Cleaner Plus is a daily cleaner for low maintenance. It both cleans and continually repairs abrasions without building gloss or leaving a film or tacky residue. Apply solution with a mop or autoscrubber. Dilute using 2 oz. with 1 gallon of water. Use in schools, offices, supermarkets, hospitals and medical centers.