Learn More
Enterprise Technology Solutions Midland Radio Corp Weather Radio, 85 dB Siren, 7"Wx2-1/2"Lx3-1/2"H, Black
SDP
Catalog No. 5000740706
Encompass_Preferred
Weather radio with an 85-decibel siren features digital S.A.M.E. technology that locks into your county's warning system to give you instant weather updates from the National Weather Service. The USB output allows you to charge devices directly from the radio. Program up to 25 counties with 80 different emergency alerts to ensure you're always aware of an emergency. Emergency power backup uses four AA batteries (sold separately) for consistent performance during a power outage. AM/FM radio with alarm buzzer allows you to listen to local stations for versatile use. Color-coded alert level indicators let you quickly see the difference between every alert.
Catalog No. 50-007-40706 Supplier Enterprise Technology Solutions Supplier No. MROWR400
Weather radio with an 85-decibel siren features digital S.A.M.E. technology that locks into your county's warning system to give you instant weather updates from the National Weather Service. The USB output allows you to charge devices directly from the radio. Program up to 25 counties with 80 different emergency alerts to ensure you're always aware of an emergency. Emergency power backup uses four AA batteries (sold separately) for consistent performance during a power outage. AM/FM radio with alarm buzzer allows you to listen to local stations for versatile use. Color-coded alert level indicators let you quickly see the difference between every alert.