Learn More
NDC Inc Midmark M11 Steam Sterilizer, 11" x 18" Chamber, Automatic Operation, 25,000 Cycles
Catalog No. NC3385938
Encompass_Preferred
The Midmark M11 Steam Sterilizer offers a robust and user-friendly solution for effective sterilization in scientific and healthcare environments. With a spacious 11" x 18" chamber, it allows for high-capacity processing of instruments in a single cycle. Its advanced steam-flush pressure pulse technology and intuitive touchscreen display differentiate it from standard models, ensuring thorough sterilization and customization options.
This item is not returnable. View return policy
Catalog No. NC3385938 Supplier NDC Inc Supplier No. M11050
Add to Cart
Add to Cart
Request a Quote
The Midmark M11 Steam Sterilizer offers a robust and user-friendly solution for effective sterilization in scientific and healthcare environments. With a spacious 11" x 18" chamber, it allows for high-capacity processing of instruments in a single cycle. Its advanced steam-flush pressure pulse technology and intuitive touchscreen display differentiate it from standard models, ensuring thorough sterilization and customization options.