Learn More
NDC Inc Midmark M9 Steam Sterilizer, 5-inch Touchscreen, 25,000 Cycle Life
Catalog No. NC3385923
Encompass_Preferred
The Midmark M9 Steam Sterilizer is a compact medical-grade autoclave designed for efficient instrument sterilization in healthcare settings. Its advanced steam pressure technology ensures effective microorganism elimination, while features like a 5-inch touchscreen display simplify operation and reduce errors. With a cycle life of 25,000 cycles and a space-optimizing design, this sterilizer enhances workflow and durability, making it a valuable asset for any medical facility.
This item is not returnable. View return policy
Catalog No. NC3385923 Supplier NDC Inc Supplier No. M9050
Add to Cart
Add to Cart
Request a Quote
The Midmark M9 Steam Sterilizer is a compact medical-grade autoclave designed for efficient instrument sterilization in healthcare settings. Its advanced steam pressure technology ensures effective microorganism elimination, while features like a 5-inch touchscreen display simplify operation and reduce errors. With a cycle life of 25,000 cycles and a space-optimizing design, this sterilizer enhances workflow and durability, making it a valuable asset for any medical facility.