Learn More
Medicom™ Midnight™ Procedure Earloop Face Mask, Black
Catalog No. 17100951 Shop All Medicom Products
Change view
Click to view available options
ASTM Level:
ASTM Level 1
ASTM Level 2
ASTM Level 3
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. ASTM Level
17-100-951 ASTM Level 3
17-100-949 ASTM Level 1
17-100-950 ASTM Level 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 17-100-951 Supplier Medicom™ Supplier No. 213992
Add to Cart
Add to Cart
Protects professionals from gaping and irritation often caused by lower-quality masks.
  • Multi-layered, shingle-pleat, earloop face mask with an adjustable nose piece.
  • Procedure masks are intended to be worn by healthcare providers to protect both the patient and the wearer from transfer of microorganisms, body fluids and particulate material.
  • Reliable protection, sophisticated style. Elegance meets protection.
  • Made from high-quality materials that increase breathability and comfort, black earloop face mask protects professionals without the gaping or irritation often caused by lower-quality masks.
  • Available in ASTM Levels 1, 2 and 3. Latex-Free.
Specifications
Product Type Medical/Procedural Face Mask
ASTM Level ASTM Level 3
Color Black
Fastening Type Earloops
Material Spunbond Polypropylene
Size One Size
Sterility Non-sterile
Style Pleated
Disposable Single-use
Latex Free Yes