Learn More
Medchemexpress LLC Midodrine-d6 hydrochloride | 1188265-43-1 | MFCD08063467 | 95.0% | 296.78 | C12H13D6ClN2O4 | 5 MG
SDP
Catalog No. 5000263977
Encompass_Preferred
Midodrine-d6 hydrochloride is the deuterated isotopologue of midodrine provided for research use as a tracer and internal standard in analytical and pharmacological studies. As an adrenergic α1-receptor agonist, it is applied in cardiovascular research and in the development and validation of quantitative analytical methods.
  • Deuterated internal standard for accurate quantification by NMR, GC-MS, and LC-MS.
  • Selective adrenergic α1-receptor agonist useful for cardiovascular research.
  • High isotopic labeling for clear mass spectrometric distinction from native analyte.
  • Available in small packaging sizes suitable for method development and standards preparation.
  • Hydrochloride salt form enhances stability and solubility for analytical workflows.
Catalog No. 50-002-63977 Supplier Medchemexpress LLC Supplier No. HY12749AS5MG
Midodrine-d6 hydrochloride is the deuterated isotopologue of midodrine provided for research use as a tracer and internal standard in analytical and pharmacological studies. As an adrenergic α1-receptor agonist, it is applied in cardiovascular research and in the development and validation of quantitative analytical methods.
  • Deuterated internal standard for accurate quantification by NMR, GC-MS, and LC-MS.
  • Selective adrenergic α1-receptor agonist useful for cardiovascular research.
  • High isotopic labeling for clear mass spectrometric distinction from native analyte.
  • Available in small packaging sizes suitable for method development and standards preparation.
  • Hydrochloride salt form enhances stability and solubility for analytical workflows.