Learn More
Medchemexpress LLC Midodrine (hydrochloride) | 43218-56-0 | 99.9% | 1 ML
SDP
Catalog No. 5000341308
Encompass_Preferred
Midodrine hydrochloride is a selective and orally active adrenergic α1-receptor agonist that strengthens vascular contraction. It is used for research on cardiovascular diseases, including orthostatic hypotension.
  • Selective and orally active adrenergic α1-receptor agonist
  • Strengthens vascular contraction
  • Used for research on cardiovascular diseases and orthostatic hypotension
  • Increases p-AMPK and PPARδ expression in C2C12, HL1, and HepG2 cells
  • Increases Ca2+ and ATP content in C2C12, HL1, and HepG2 cells
  • Reduces cellular lipid content in 3T3-L1 cells
  • Reduces platelet-activating factor (Paf)-induced death in mice
  • Demonstrates anti-hypotensive effects in orthostatic hypotension rat models
  • Relieves hypertension in spontaneously hypertensive rat models
Catalog No. 50-003-41308 Supplier Medchemexpress LLC Supplier No. HY12749A10MM/1ML
Midodrine hydrochloride is a selective and orally active adrenergic α1-receptor agonist that strengthens vascular contraction. It is used for research on cardiovascular diseases, including orthostatic hypotension.
  • Selective and orally active adrenergic α1-receptor agonist
  • Strengthens vascular contraction
  • Used for research on cardiovascular diseases and orthostatic hypotension
  • Increases p-AMPK and PPARδ expression in C2C12, HL1, and HepG2 cells
  • Increases Ca2+ and ATP content in C2C12, HL1, and HepG2 cells
  • Reduces cellular lipid content in 3T3-L1 cells
  • Reduces platelet-activating factor (Paf)-induced death in mice
  • Demonstrates anti-hypotensive effects in orthostatic hypotension rat models
  • Relieves hypertension in spontaneously hypertensive rat models