Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Midodrine hydrochloride | 43218-56-0 | 99.9% | 10 MG
SDP

Catalog No. 5000338467
Encompass_Preferred

Midodrine hydrochloride is a selective and orally active adrenergic α1-receptor agonist. It strengthens vascular contraction and is utilized in research concerning cardiovascular diseases, such as orthostatic hypotension.

  • Selective and orally active adrenergic α1-receptor agonist
  • Strengthens vascular contraction
  • Utilized in research for cardiovascular diseases like orthostatic hypotension
  • Increases p-AMPK and PPARδ expression
  • Increases Ca2+ in C2C12, HL1 and HepG2 cells
  • Increases maximal mitochondrial oxidative phosphorylation and ATP content in C2C12 cells
  • Reduces cellular lipid content in 3T3-L1 cells
  • Reduces platelet-activating factor-induced death in mice
  • Shows anti-hypotensive effects in orthostatic hypotension rat models
  • Relieves hypertension in spontaneously hypertensive rat models

Catalog No. 50-003-38467 Supplier Medchemexpress LLC Supplier No. HY12749A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Midodrine hydrochloride is a selective and orally active adrenergic α1-receptor agonist. It strengthens vascular contraction and is utilized in research concerning cardiovascular diseases, such as orthostatic hypotension.

  • Selective and orally active adrenergic α1-receptor agonist
  • Strengthens vascular contraction
  • Utilized in research for cardiovascular diseases like orthostatic hypotension
  • Increases p-AMPK and PPARδ expression
  • Increases Ca2+ in C2C12, HL1 and HepG2 cells
  • Increases maximal mitochondrial oxidative phosphorylation and ATP content in C2C12 cells
  • Reduces cellular lipid content in 3T3-L1 cells
  • Reduces platelet-activating factor-induced death in mice
  • Shows anti-hypotensive effects in orthostatic hypotension rat models
  • Relieves hypertension in spontaneously hypertensive rat models

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.