Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Midodrine (hydrochloride) | 43218-56-0 | 99.9% | 25 MG
SDP

Catalog No. 5000341869
Encompass_Preferred

Midodrine hydrochloride is a selective and orally active adrenergic α1-receptor agonist. It strengthens vascular contraction and can be used for research on cardiovascular diseases such as orthostatic hypotension.

  • Strengthens vascular contraction
  • Increases p-AMPK and PPARδ expression in C2C12, HL1, and HepG2 cells
  • Increases Ca2+ in C2C12, HL1, and HepG2 cells
  • Increases maximal mitochondrial oxidative phosphorylation and ATP content in C2C12 cells
  • Reduces cellular lipid content in 3T3-L1 cells
  • Reduces platelet-activating factor (Paf)-induced death in mice
  • Shows anti-hypotensive effects in orthostatic hypotension rat models
  • Relieves hypertension in spontaneously hypertensive rat models

Catalog No. 50-003-41869 Supplier Medchemexpress LLC Supplier No. 25MGHY12749A
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Midodrine hydrochloride is a selective and orally active adrenergic α1-receptor agonist. It strengthens vascular contraction and can be used for research on cardiovascular diseases such as orthostatic hypotension.

  • Strengthens vascular contraction
  • Increases p-AMPK and PPARδ expression in C2C12, HL1, and HepG2 cells
  • Increases Ca2+ in C2C12, HL1, and HepG2 cells
  • Increases maximal mitochondrial oxidative phosphorylation and ATP content in C2C12 cells
  • Reduces cellular lipid content in 3T3-L1 cells
  • Reduces platelet-activating factor (Paf)-induced death in mice
  • Shows anti-hypotensive effects in orthostatic hypotension rat models
  • Relieves hypertension in spontaneously hypertensive rat models

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.