Learn More
Medchemexpress LLC HY-10230 10mg , Midostaurin CAS:120685-11-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1023010MG
Encompass_Preferred
HY-10230 10mg Midostaurin CAS: 120685-11-2 Midostaurin (PKC412; CGP 41251) is an orally active, reversible multi-targeted protein kinase inhibitor. Midostaurin inhibits PKCα/β/γ, Syk, Flk-1, Akt, PKA, c-Kit, c-Fgr, c-Src, FLT3, PDFRβ and VEGFR1/2 with IC50s ranging from 22-500 nM. Midostaurin also upregulates endothelial nitric oxide synthase (eNOS) gene expression. Midostaurin shows powerful anticancer effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9603