Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Midostaurin-d5 | 00-00-0 | 98.0% | 575.67 g/mol | C35H25D5N4O4 | 1 MG
SDP

Catalog No. 5000247443
Encompass_Preferred

Midostaurin-d5 is a deuterium-labeled analogue of midostaurin supplied as a powder for analytical and biochemical research. It is intended primarily as an isotopically labeled internal standard for pharmacokinetic, metabolic, and bioanalytical assays.

  • Deuterium-labeled internal standard for midostaurin studies.
  • Molecular formula C35H25D5N4O4; molecular weight 575.67 g/mol.
  • Purity 98.0% as supplied.
  • Physical form: powder; typical package size 1 mg.
  • Storage: powder -20°C (up to 3 years) or 4°C (up to 2 years).
  • Solubility: DMSO ≈15 mg/mL; sonication and warming recommended.

Catalog No. 50-002-47443 Supplier Medchemexpress LLC Supplier No. HY10230S1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Midostaurin-d5 is a deuterium-labeled analogue of midostaurin supplied as a powder for analytical and biochemical research. It is intended primarily as an isotopically labeled internal standard for pharmacokinetic, metabolic, and bioanalytical assays.

  • Deuterium-labeled internal standard for midostaurin studies.
  • Molecular formula C35H25D5N4O4; molecular weight 575.67 g/mol.
  • Purity 98.0% as supplied.
  • Physical form: powder; typical package size 1 mg.
  • Storage: powder -20°C (up to 3 years) or 4°C (up to 2 years).
  • Solubility: DMSO ≈15 mg/mL; sonication and warming recommended.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.