Learn More
Selleck Chemical LLC Midostaurin (PKC412) 5mg 120685-11-2 CGP 41251
SDP
Catalog No. 501365621
Encompass_Preferred
Midostaurin (pkc412, CGP 41251) is a multi-targeted kinase inhibitor, including PKC?/?/?, Syk, Flk-1, Akt, PKA, c-Kit, c-Fgr, c-Src, FLT3, PDFR? and VEGFR1/2 with IC50 ranging from 80-500 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5621 Supplier Selleck Chemical LLC Supplier No. S80645MG
Midostaurin (pkc412, CGP 41251) is a multi-targeted kinase inhibitor, including PKC?/?/?, Syk, Flk-1, Akt, PKA, c-Kit, c-Fgr, c-Src, FLT3, PDFR? and VEGFR1/2 with IC50 ranging from 80-500 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.