Learn More
Midwest Scientific MIDSCI Pipette Controller with retractable stand
SDP
Catalog No. NC1124300
Encompass
MIDSCI Pipette Controllers are lightweight (only 6.3oz/170g) and designed to fit comfortably in either the right or the left hand. The natural feel of the ergonomic body design, the computer balanced weight distribution and the low pressure fingertip control buttons...all combine to provide a more comfortable pipetting experience. With QuickStand Retractable Stand; Variable Speed Control; Cordless and rechargeable; 8 hours of continuous operation; For pipettes from 1ml to 100ml, 2 year warranty
Catalog No. NC1124300 Supplier Midwest Scientific Supplier No. MPIP1
MIDSCI Pipette Controllers are lightweight (only 6.3oz/170g) and designed to fit comfortably in either the right or the left hand. The natural feel of the ergonomic body design, the computer balanced weight distribution and the low pressure fingertip control buttons...all combine to provide a more comfortable pipetting experience. With QuickStand Retractable Stand; Variable Speed Control; Cordless and rechargeable; 8 hours of continuous operation; For pipettes from 1ml to 100ml, 2 year warranty