Learn More
National Safety Apparel™ Midweight Double-Layer FR Knit Hood
SDP
Catalog No. 17001709 Shop All National Safety Apparel Products
Encompass_Preferred
Change view
Click to view available options
Color: White
Catalog No.
17-001-709 Color: White
1 options
Catalog No. 17-001-709 Supplier National Safety Apparel™ Supplier No. H61MH
Double layer hood, one size fits most
  • Made from 6 oz.
  • Modacrylic, DuPont™ Nomex™ Blend
  • Mid-weight
  • Double layer hood
Specifications
Size Medium
Material DuPont™ Nomex™, Modacrylic Blend
Color White
Seam Type Stitched
Arc Flash Rating 19 cal/cm²
Certifications/Compliance CAT 2, NFPA 70 E, ASTM F1506, CSA Z462, OSHA 1910.269