Learn More
Aviva Systems Biology Corp MIER1 Antibody - middle region (ARP34468_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34468_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against MIER1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MIER1. Alternate names: ER1, MI-ER1, hMI-ER1, RP5-944N15.1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55406