Learn More
Aviva Systems Biology Corp MIER3 Antibody - middle region (ARP39920_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39920_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MIER3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MIER3. Alternate names: DKFZP781I1119, DKFZp686L09111, DKFZp781G1119, DKFZp781I1119, FLJ35954ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast
Catalog No. 50-014-57232