Learn More
Aviva Systems Biology Corp MIF Antibody - middle region (ARP56347_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56347_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MIF. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human MIF. Alternate names: GIF, GLIF, MMIFApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-286-141