Learn More
ACROBiosystems ACROBiosystems Biotinylated Human MIF Protein, His,Avitag™
Supplier:  ACROBiosystems MIFH82E7200UG
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human MIF, His,Avitag (MIF-H82E7) is expressed from human 293 cells (HEK293). It contains AA Pro 2 - Ala 115 (Accession # P14174-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 16.0 kDa. The protein migrates as 20 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-6968