Learn More
Genscript Corporation Antibody Protein Proteins Proteins 10 UG 1/EA
Supplier:  Genscript Corporation Z0315910
Encompass
Macrophage Migration Inhibitory Factor (MIF) is a pleiotropic cytokine existing as a homotrimer in vivo. MIF was originally identified as a T cell derived factor responsible for the inhibition of macrophage migration. However recently MIF has received much more attention because of its possible roles in angiogenesis and cancer development. MIF is over-expressed in various cancers including pancreatic breast colon brain prostate skin and lung. The intratumoral expression of MIF is strongly correlated with angiogenic growth factor expression such as the expression of Interleukin 8 (IL-8) and Vascular Endothelial Growth Factor (VEGF) and with risk of recurrence after resection.
Catalog No. 50-252-9766