Learn More
Medchemexpress LLC MIF protein, human (HEK293, His) | >95.0% | 50UG
SDP
Catalog No. 5000177086
Encompass_Preferred
Recombinant human macrophage migration inhibitory factor (MIF) expressed in HEK293 mammalian cells with a C-terminal His tag. Supplied lyophilized from PBS (pH 7.4) with 8% trehalose, the protein is provided at high purity and is suitable for biochemical and cell-based assays. Predicted molecular mass is 13.4 kDa; observed ~25-30 kDa on SDS-PAGE due to glycosylation.
  • Expressed in HEK293 mammalian cells with a C-terminal His tag.
  • Lyophilized formulation in PBS, pH 7.4, containing 8% trehalose.
  • High purity (>95%) as determined by Tris-Bis PAGE.
  • Predicted molecular mass 13.4 kDa; apparent ~25-30 kDa on SDS-PAGE (glycosylated).
  • Suitable for biochemical and cell-based assays.
Catalog No. 50-001-77086 Supplier Medchemexpress LLC Supplier No. HYP70079250UG
Recombinant human macrophage migration inhibitory factor (MIF) expressed in HEK293 mammalian cells with a C-terminal His tag. Supplied lyophilized from PBS (pH 7.4) with 8% trehalose, the protein is provided at high purity and is suitable for biochemical and cell-based assays. Predicted molecular mass is 13.4 kDa; observed ~25-30 kDa on SDS-PAGE due to glycosylation.
  • Expressed in HEK293 mammalian cells with a C-terminal His tag.
  • Lyophilized formulation in PBS, pH 7.4, containing 8% trehalose.
  • High purity (>95%) as determined by Tris-Bis PAGE.
  • Predicted molecular mass 13.4 kDa; apparent ~25-30 kDa on SDS-PAGE (glycosylated).
  • Suitable for biochemical and cell-based assays.