Learn More
Bachem Melanocyte-Stimulating Hormone-Release Inhibiting Factor
SDP
Supplier:  Bachem 4000343.0005
Encompass_Preferred
Melanocyte-Stimulating Hormone-Release Inhibiting Factor, H-Pro-Leu-Gly-NH2, 2002-44-0, C13H24N4O3, Mr 284.36, 5g. Melanostatin, MIF-I, Melanotropin Release-Inhibiting Factor, MSH Release Inhibiting Factor, amide. MIF-I, the C-terminal tripeptide of oxytocin, has been shown to be active in numerous behavioral tests and clinical situations.
Catalog No. 50-194-6870