Learn More
Medchemexpress LLC HY-P7388 50ug Medchemexpress, MIF, Mouse CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP738850UG
Encompass_Preferred
Medchemexpress, HY-P7388 50ug MIF, Mouse CAS: Recombinant Mouse Macrophage Migration Inhibitory Factor has assumed an important role as a pivotal regulator of innate immunity. Recombinant Mouse Macrophage Migration Inhibitory Factor promotes the pro-inflammatory functions of immune cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4386