Learn More
Medchemexpress LLC Macrophage migration inhibitory factor (mouse) | 97.0% | 20UG
SDP
Catalog No. 5000175462
Encompass_Preferred
Recombinant mouse macrophage migration inhibitory factor (MIF) expressed in Pichia pastoris with an N-terminal 6xHis tag. Supplied lyophilized for research use, this protein supports studies of innate immunity, inflammation, enzymatic activity, and receptor interactions, and is formulated for low endotoxin and long-term storage.
  • High purity (>97% by reducing SDS-PAGE)
  • Low endotoxin (<1 EU/μg) suitable for cell-based assays
  • Expressed in Pichia pastoris with N-terminal 6xHis tag
  • Lyophilized formulation for long-term storage and stability
  • Stable at -20°C; reconstituted stability at 4°C for 1 week
  • Includes sequence and UniProt accession P34884 for traceability
Catalog No. 50-001-75462 Supplier Medchemexpress LLC Supplier No. HYP70051720UG
Recombinant mouse macrophage migration inhibitory factor (MIF) expressed in Pichia pastoris with an N-terminal 6xHis tag. Supplied lyophilized for research use, this protein supports studies of innate immunity, inflammation, enzymatic activity, and receptor interactions, and is formulated for low endotoxin and long-term storage.
  • High purity (>97% by reducing SDS-PAGE)
  • Low endotoxin (<1 EU/μg) suitable for cell-based assays
  • Expressed in Pichia pastoris with N-terminal 6xHis tag
  • Lyophilized formulation for long-term storage and stability
  • Stable at -20°C; reconstituted stability at 4°C for 1 week
  • Includes sequence and UniProt accession P34884 for traceability