Learn More
Aviva Systems Biology Corp MIF4GD Antibody - C-terminal region (ARP40950_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40950_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against MIF4GD. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human MIF4GD. Alternate names: MIFD, AD023, SLIP1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53253