Learn More
Medchemexpress LLC HY-13682 5mg , Mifamurtide CAS:83461-56-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY136825MG
Encompass_Preferred
HY-13682 5mg Mifamurtide CAS: 83461-56-7 Mifamurtide (MTP-PE), an analog of the muramyl dipeptide (MDP), is a nonspecific immunomodulator by stimulating the immune response activating macrophages and monocytes. Mifamurtide, an orphan drug, is a specific ligand of NOD2 used as an insulin sensitizer. Mifamurtide has the potential for osteosarcoma research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1410