Learn More
Medchemexpress LLC Mifamurtide sodium | 90825-43-7 | 98.19% | 1259.48 | 1 ML
SDP
Catalog No. 5000348977
Encompass_Preferred
Mifamurtide sodium is an analog of the muramyl dipeptide (MDP) that functions as a nonspecific immunomodulator. It stimulates the immune response by activating macrophages and monocytes, acting as a specific ligand for NOD2 and an insulin sensitizer. This compound shows potential for use in rare disease and osteosarcoma research.
  • Functions as a nonspecific immunomodulator
  • Activates macrophages and monocytes
  • Acts as a specific ligand for NOD2
  • Serves as an insulin sensitizer
  • Useful in rare disease and osteosarcoma research
  • Increases tumoricidal activity
  • Promotes secretion of pro-inflammatory cytokines
Catalog No. 50-003-48977 Supplier Medchemexpress LLC Supplier No. HY13682B10MM/1ML
Mifamurtide sodium is an analog of the muramyl dipeptide (MDP) that functions as a nonspecific immunomodulator. It stimulates the immune response by activating macrophages and monocytes, acting as a specific ligand for NOD2 and an insulin sensitizer. This compound shows potential for use in rare disease and osteosarcoma research.
  • Functions as a nonspecific immunomodulator
  • Activates macrophages and monocytes
  • Acts as a specific ligand for NOD2
  • Serves as an insulin sensitizer
  • Useful in rare disease and osteosarcoma research
  • Increases tumoricidal activity
  • Promotes secretion of pro-inflammatory cytokines