Learn More
Medchemexpress LLC Mifamurtide (TFA) | 98.0% | 1351.52 | 1 MG
SDP
Catalog No. 5000365248
Encompass_Preferred
Mifamurtide TFA (MTP-PE TFA) is an analog of the muramyl dipeptide (MDP) that functions as a nonspecific immunomodulator by stimulating the immune response, activating macrophages and monocytes. It is a specific ligand for NOD2 and acts as an insulin sensitizer, with potential applications in rare disease and osteosarcoma research.
  • Induces reduction of MG63 cell numbers when co-cultured with macrophages.
  • Increases M1 polarization marker iNOS and M2 polarization marker CD206 mRNAs.
  • Increases pro-inflammatory (IL-1β, IL-6) and anti-inflammatory (IL-4, IL-10) cytokines.
  • Activates macrophages and monocytes, leading to upregulation of tumoricidal activity and secretion of pro-inflammatory cytokines.
  • Shows a trend of diminished spontaneous lung metastasis dissemination in osteosarcoma models.
  • Improves glucose tolerance during endotoxemia and in HFD-fed mice.
Catalog No. 50-003-65248 Supplier Medchemexpress LLC Supplier No. 1MGHY13682C
Mifamurtide TFA (MTP-PE TFA) is an analog of the muramyl dipeptide (MDP) that functions as a nonspecific immunomodulator by stimulating the immune response, activating macrophages and monocytes. It is a specific ligand for NOD2 and acts as an insulin sensitizer, with potential applications in rare disease and osteosarcoma research.
  • Induces reduction of MG63 cell numbers when co-cultured with macrophages.
  • Increases M1 polarization marker iNOS and M2 polarization marker CD206 mRNAs.
  • Increases pro-inflammatory (IL-1β, IL-6) and anti-inflammatory (IL-4, IL-10) cytokines.
  • Activates macrophages and monocytes, leading to upregulation of tumoricidal activity and secretion of pro-inflammatory cytokines.
  • Shows a trend of diminished spontaneous lung metastasis dissemination in osteosarcoma models.
  • Improves glucose tolerance during endotoxemia and in HFD-fed mice.