Learn More
Apexbio Technology LLC Mifepristone 84371-65-3 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062033
Encompass_Preferred
Mifepristone (RU486) is a synthetic steroid compound functioning predominantly as a progesterone receptor antagonist By competing with progesterone for receptor binding it disrupts progesterone-mediated signaling pathways and alters gene expression involved in cellular proliferation and differentiation Research applications of mifepristone encompass its use in reproductive biology studies as an antagonist to progesterone-driven processes Additionally due to its modulatory effects on cellular proliferation pathways mifepristone is investigated in oncology studies to understand potential antiproliferative effects in various cancer models including uterine fibroids meningiomas breast prostate endometrial gastric and ovarian cancer cell lines where it demonstrates inhibitory activity in a dose-dependent manner
Catalog No. 50-000-62033 Supplier Apexbio Technology LLC Supplier No. B15115.1
Mifepristone (RU486) is a synthetic steroid compound functioning predominantly as a progesterone receptor antagonist By competing with progesterone for receptor binding it disrupts progesterone-mediated signaling pathways and alters gene expression involved in cellular proliferation and differentiation Research applications of mifepristone encompass its use in reproductive biology studies as an antagonist to progesterone-driven processes Additionally due to its modulatory effects on cellular proliferation pathways mifepristone is investigated in oncology studies to understand potential antiproliferative effects in various cancer models including uterine fibroids meningiomas breast prostate endometrial gastric and ovarian cancer cell lines where it demonstrates inhibitory activity in a dose-dependent manner