Learn More
Medchemexpress LLC HY-13683 500mg , Mifepristone RU486;RU 38486 CAS:84371-65-3 Purity:98%
SDP
Catalog No. 501492370
Encompass_Preferred
Medchemexpress, HY-13683 500mg Mifepristone RU486;RU 38486 CAS:84371-65-3 Formula:C29H35NO2 IC50: 0.2 nM (progesterone receptor, in T47D cells), 2.6 nM (glucocorticoid receptor, in A549 cells) Purity:98% Mifepristone is a progesterone receptor ( PR ) antagonist ( IC 50 =0.2 nM) in a T47D cell-based assay, also is a glucocorticoid receptor ( GR ) antagonist ( IC 50 =2.6 nM) in an A549 cell-based assay. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2370 Supplier Medchemexpress LLC Supplier No. HY13683500MG
Medchemexpress, HY-13683 500mg Mifepristone RU486;RU 38486 CAS:84371-65-3 Formula:C29H35NO2 IC50: 0.2 nM (progesterone receptor, in T47D cells), 2.6 nM (glucocorticoid receptor, in A549 cells) Purity:98% Mifepristone is a progesterone receptor ( PR ) antagonist ( IC 50 =0.2 nM) in a T47D cell-based assay, also is a glucocorticoid receptor ( GR ) antagonist ( IC 50 =2.6 nM) in an A549 cell-based assay. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.