Learn More
Selleck Chemical LLC Mifepristone 50mg 84371-65-3 RU486, C-1073
SDP
Catalog No. 501362622
Encompass_Preferred
Mifepristone (RU486, C-1073) is a remarkably active antagonist of progesterone receptor and glucocorticoid receptor with IC50 of 0.2 nM and 2.6 nM, respectively. Mifepristone promotes cell autophagy and apoptosis, decreases Bcl-2 level and increases Beclin1 level, accompanied by weakened interaction between Bcl-2 and Beclin1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-2622 Supplier Selleck Chemical LLC Supplier No. S260650MG
Mifepristone (RU486, C-1073) is a remarkably active antagonist of progesterone receptor and glucocorticoid receptor with IC50 of 0.2 nM and 2.6 nM, respectively. Mifepristone promotes cell autophagy and apoptosis, decreases Bcl-2 level and increases Beclin1 level, accompanied by weakened interaction between Bcl-2 and Beclin1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.