Learn More
Medchemexpress LLC Mifepristone | 84371-65-3 | 99.9% | 429.59 | 1 ML
SDP
Catalog No. 5000348693
Encompass_Preferred
Mifepristone (RU486) is a progesterone receptor (PR) and glucocorticoid receptor (GR) antagonist with IC50s of 0.2 nM and 2.6 nM in in vitro assay. It was discovered as the first competitive progesterone antagonist, stimulating a search for more potent and selective antiprogestins.
  • Progesterone receptor and glucocorticoid receptor antagonist
  • Inhibits cell growth
  • Potentiates doxorubicin-induced cytotoxicity in K562/R7 cells
  • Reduces severity of EtOH withdrawal in vivo
Catalog No. 50-003-48693 Supplier Medchemexpress LLC Supplier No. HY1368310MM/1ML
Mifepristone (RU486) is a progesterone receptor (PR) and glucocorticoid receptor (GR) antagonist with IC50s of 0.2 nM and 2.6 nM in in vitro assay. It was discovered as the first competitive progesterone antagonist, stimulating a search for more potent and selective antiprogestins.
  • Progesterone receptor and glucocorticoid receptor antagonist
  • Inhibits cell growth
  • Potentiates doxorubicin-induced cytotoxicity in K562/R7 cells
  • Reduces severity of EtOH withdrawal in vivo