Learn More
Medchemexpress LLC Mifepristone-d3 | 1228097-18-4 | 97.7% | 432.61 | 1 MG
SDP
Catalog No. 5000348577
Encompass_Preferred
Mifepristone-d3 is the deuterium labeled Mifepristone. It acts as a progesterone receptor (PR) and glucocorticoid receptor (GR) antagonist with IC50s of 0.2 nM and 2.6 nM in in vitro assay. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium labeled compound
  • Acts as a progesterone receptor antagonist
  • Acts as a glucocorticoid receptor antagonist
  • Useful as a tracer in drug development processes
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-48577 Supplier Medchemexpress LLC Supplier No. HY13683S1MG
Mifepristone-d3 is the deuterium labeled Mifepristone. It acts as a progesterone receptor (PR) and glucocorticoid receptor (GR) antagonist with IC50s of 0.2 nM and 2.6 nM in in vitro assay. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium labeled compound
  • Acts as a progesterone receptor antagonist
  • Acts as a glucocorticoid receptor antagonist
  • Useful as a tracer in drug development processes
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS