Learn More
Medchemexpress LLC Mifepristone (standard) | 84371-65-3 | 99.4% | 429.59 | 100 MG
SDP
Catalog No. 5000348976
Encompass_Preferred
Mifepristone (Standard) is an analytical standard of Mifepristone, also known as RU486. It functions as an antagonist for progesterone receptor (PR) and glucocorticoid receptor (GR), exhibiting IC50s of 0.2 nM and 2.6 nM respectively in in vitro assays. This product is designed for research and analytical applications.
  • Intended for research and analytical applications
  • Functions as a progesterone receptor (PR) and glucocorticoid receptor (GR) antagonist
  • Used in qualitative, quantitative, and methodological research experiments
  • Suitable for techniques such as HPLC, GC, and MS
  • Serves as a reference standard supplied assay
Catalog No. 50-003-48976 Supplier Medchemexpress LLC Supplier No. HY13683R100MG
Mifepristone (Standard) is an analytical standard of Mifepristone, also known as RU486. It functions as an antagonist for progesterone receptor (PR) and glucocorticoid receptor (GR), exhibiting IC50s of 0.2 nM and 2.6 nM respectively in in vitro assays. This product is designed for research and analytical applications.
  • Intended for research and analytical applications
  • Functions as a progesterone receptor (PR) and glucocorticoid receptor (GR) antagonist
  • Used in qualitative, quantitative, and methodological research experiments
  • Suitable for techniques such as HPLC, GC, and MS
  • Serves as a reference standard supplied assay