Learn More
Cayman Chemical Mifepristoned3 GC MS Standards
SDP
Catalog No. 502890387
Encompass_Preferred
An internal standard for the quantification of mifepristone by GC- or LC-MS
Catalog No. 50-289-0387 Supplier Cayman Chemical Supplier No. 100106601 MG
An internal standard for the quantification of mifepristone by GC- or LC-MS