Learn More
CELLULAR TECHNOLOGY LTD MIFNGP-2M/2, ELISPOT KIT
SDP
Catalog No. NC2024984
Encompass
mIFNgp-2M/2, Mouse IFN-g Single-Color ELISPOT Kit of 2 pre-coated 96-well Strip plates
Catalog No. NC2024984 Supplier CELLULAR TECHNOLOGY LTD Supplier No. MIFNGP2M/2
mIFNgp-2M/2, Mouse IFN-g Single-Color ELISPOT Kit of 2 pre-coated 96-well Strip plates