Learn More
Medchemexpress LLC HY-100277 5mg Medchemexpress, Mifobate CAS:76541-72-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1002775MG
Encompass_Preferred
Medchemexpress, HY-100277 5mg Mifobate CAS:76541-72-5 Mifobate (SR-202) is a potent and specific PPARγ antagonist. Mifobate (SR-202) selectively inhibits Thiazolidinedione (TZD)-induced PPARγ transcriptional activity (IC50=140 μM). Mifobate (SR-202) does not affect basal or ligand-stimulated transcriptional activity of PPARα, PPARβ, or the farnesoid X receptor (FXR). Mifobate (SR-202) shows antiobesity and antidiabetic effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7722