Learn More
Medchemexpress LLC HY-P7253 10ug Medchemexpress, MIG/CXCL9, Human CAS: Purity:>98%
SDP
Catalog No. 501970588
Encompass_Preferred
Medchemexpress, HY-P7253 10ug MIG/CXCL9, Human CAS: Recombinant Human MIG/CXCL9 is a chemokine binding to CXCR3, associated with tumor progression, immune escape, and angiogenesis in several malignancies. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0588 Supplier Medchemexpress LLC Supplier No. HYP725310UG
Medchemexpress, HY-P7253 10ug MIG/CXCL9, Human CAS: Recombinant Human MIG/CXCL9 is a chemokine binding to CXCR3, associated with tumor progression, immune escape, and angiogenesis in several malignancies. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.