Learn More
Medchemexpress LLC MIG/CXCL9 Human HE 20ug
SDP
Catalog No. 5000182384
Encompass_Preferred
CXCL9 also known as MIG is one member of the ELR-negative CXC chemokine subfamily and can be induced by IFN- CXCL9 binds to its receptor CXCR3 and can recruit CXCR3 cells such as effector T cells regulatory T cells (Tregs) and CD8 cytotoxic T cells CXCL9 is involved in immunoregulatory and inflammatory processes but it also play a key role in tumor growth angiogenesis and metastasis[1][2] MIG/CXCL9 Protein Human (HEK293 His) is produced in HEK293 cells with six C-Terminal His-tags It consists of 103 amino acids (T23-T125)
Catalog No. 50-001-82384 Supplier Medchemexpress LLC Supplier No. HYP7267520UG
CXCL9 also known as MIG is one member of the ELR-negative CXC chemokine subfamily and can be induced by IFN- CXCL9 binds to its receptor CXCR3 and can recruit CXCR3 cells such as effector T cells regulatory T cells (Tregs) and CD8 cytotoxic T cells CXCL9 is involved in immunoregulatory and inflammatory processes but it also play a key role in tumor growth angiogenesis and metastasis[1][2] MIG/CXCL9 Protein Human (HEK293 His) is produced in HEK293 cells with six C-Terminal His-tags It consists of 103 amino acids (T23-T125)