Learn More
Medchemexpress LLC HY-14929A 5mg Medchemexpress, Migalastat (hydrochloride) CAS:75172-81-5 Purity:>98%
SDP
Catalog No. 502081660
Encompass_Preferred
Medchemexpress, HY-14929A 5mg Migalastat (hydrochloride) CAS:75172-81-5 Migalastat hydrochloride (GR181413A) is a potent and competitive inhibitor of α-galactosidase A (α-Gal A) with an IC50 of 0.04 μM for human α-Gal A. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1660 Supplier Medchemexpress LLC Supplier No. HY14929A5MG
Medchemexpress, HY-14929A 5mg Migalastat (hydrochloride) CAS:75172-81-5 Migalastat hydrochloride (GR181413A) is a potent and competitive inhibitor of α-galactosidase A (α-Gal A) with an IC50 of 0.04 μM for human α-Gal A. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.