Learn More
Hoefer, Inc Hoefer™ SE260 Mighty Small II Deluxe Mini Vertical Protein Electrophoresis Unit (Complete unit with .75mm)
SDP
Catalog No. NC2740675
Encompass_Preferred
Combines the advantages of small format and efficient cooling for rapid screening of proteins and nucleic acidsFeatures:Maximum glass plate size 10 x 10.5 cmFlexible design, additionally able to accommodate 10 x 8 cm format with optional accessoriesSupports a wide variety of precast gelsEfficient active cooling ensures sharp bandsRun up to two gels (maximum 30 samples) at one time under identical conditionsQuick and easy to assembleComplete Units Include:Buffer Chamber/Cooling Core; Safety Lid with High Voltage Leads; Glass Plates, Rectangular, 10 x 10.5 cm, 10 pcs; Alumina Plates, Notched, 10 x 10.5 cm, 5 pcs; Combs, 10 well, 2 pcs; T-Spacers, 4 pcs; Gel casterBasic Unit Includes:The same as complete unit without the combs, spacers, plates, and caster. Specifications:Glass Plate Size (W x H) 10 x 10.5 cm or 10 x 8 cmSpacers 0.75 mm, 1.0 mm, 1.5 mmMaximum Power Settings 500 V, 500 mA, 12 WUnit Dimensions (W x H x D) 16.5 x 18 x 16 cm
Catalog No. NC2740675 Supplier Hoefer, Inc Supplier No. SE26010A.75
Combines the advantages of small format and efficient cooling for rapid screening of proteins and nucleic acidsFeatures:Maximum glass plate size 10 x 10.5 cmFlexible design, additionally able to accommodate 10 x 8 cm format with optional accessoriesSupports a wide variety of precast gelsEfficient active cooling ensures sharp bandsRun up to two gels (maximum 30 samples) at one time under identical conditionsQuick and easy to assembleComplete Units Include:Buffer Chamber/Cooling Core; Safety Lid with High Voltage Leads; Glass Plates, Rectangular, 10 x 10.5 cm, 10 pcs; Alumina Plates, Notched, 10 x 10.5 cm, 5 pcs; Combs, 10 well, 2 pcs; T-Spacers, 4 pcs; Gel casterBasic Unit Includes:The same as complete unit without the combs, spacers, plates, and caster. Specifications:Glass Plate Size (W x H) 10 x 10.5 cm or 10 x 8 cmSpacers 0.75 mm, 1.0 mm, 1.5 mmMaximum Power Settings 500 V, 500 mA, 12 WUnit Dimensions (W x H x D) 16.5 x 18 x 16 cm