Learn More
Abcam Miglustat hydrochloride, 1MG
Catalog No. 502671225
Encompass_Preferred
MW 255.74 Da, Purity >=98%. An inhibitor of the N-linked oligosaccharide processing enzymes α-glucosidase I and II. It inhibits rat testicular enzymes ceramide-glucosyltransferase and beta-glucosidase 2 with IC₅₀ values of 32 and 81 μM respectively. Miglustat is used for the treatment of type 1 Gaucher's disease. It is an inhibitor of HIV replication in vitro. It shows activity against SARS-CoV-2 in vitro with EC₅₀ values in the micromolar range.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-1225 Supplier Abcam Supplier No. AB2869361MG
MW 255.74 Da, Purity >=98%. An inhibitor of the N-linked oligosaccharide processing enzymes α-glucosidase I and II. It inhibits rat testicular enzymes ceramide-glucosyltransferase and beta-glucosidase 2 with IC₅₀ values of 32 and 81 μM respectively. Miglustat is used for the treatment of type 1 Gaucher's disease. It is an inhibitor of HIV replication in vitro. It shows activity against SARS-CoV-2 in vitro with EC₅₀ values in the micromolar range.
The product is subject to the following: Abcam Restricted Use Statement