Learn More
Medchemexpress LLC Miglustat hydrochloride | 210110-90-0 | 98.0% | 255.74 | 100 MG
SDP
Catalog No. 5000384743
Encompass_Preferred
Miglustat hydrochloride is an orally active, reversible ceramide glucosyltransferase inhibitor with blood-brain barrier permeability. It is primarily used for the research of type I Gaucher disease. It also restores F508del-CFTR function in cystic fibrosis (CF) bronchial epithelial cells and reduces the inflammatory response to P. aeruginosa.
  • Orally active
  • Reversible ceramide glucosyltransferase inhibitor
  • Crosses the blood-brain barrier
  • Used in research for type I Gaucher disease
  • Restores F508del-CFTR function in cystic fibrosis bronchial epithelial cells
  • Reduces inflammatory response to P. aeruginosa
Catalog No. 50-003-84743 Supplier Medchemexpress LLC Supplier No. HY17020A100MG
Miglustat hydrochloride is an orally active, reversible ceramide glucosyltransferase inhibitor with blood-brain barrier permeability. It is primarily used for the research of type I Gaucher disease. It also restores F508del-CFTR function in cystic fibrosis (CF) bronchial epithelial cells and reduces the inflammatory response to P. aeruginosa.
  • Orally active
  • Reversible ceramide glucosyltransferase inhibitor
  • Crosses the blood-brain barrier
  • Used in research for type I Gaucher disease
  • Restores F508del-CFTR function in cystic fibrosis bronchial epithelial cells
  • Reduces inflammatory response to P. aeruginosa