Learn More
Medchemexpress LLC Miglustat hydrochloride | 210110-90-0 | 98.0% | 255.74 | 50 MG
SDP
Catalog No. 5000385861
Encompass_Preferred
Miglustat (N-Butyldeoxynojirimycin) hydrochloride is an orally active and reversible ceramide glucosyltransferase inhibitor, with blood-brain barrier permeability. It can be used for the research of type I gaucher disease.
  • Orally active and reversible ceramide glucosyltransferase inhibitor
  • Blood-brain barrier permeability
  • Used for research of type I Gaucher disease
  • Restores F508del-CFTR function in cystic fibrosis (CF) bronchial epithelial IB3-1 and CuFi-1 cells
  • Reduces inflammatory response to P. aeruginosa in both CF and non-CF bronchial cells
  • Able to rescue synaptic plasticity deficits, to restore ERKs activation and to counteract hyperexcitability in NPC1-/- mice
Catalog No. 50-003-85861 Supplier Medchemexpress LLC Supplier No. HY17020A50MG
Miglustat (N-Butyldeoxynojirimycin) hydrochloride is an orally active and reversible ceramide glucosyltransferase inhibitor, with blood-brain barrier permeability. It can be used for the research of type I gaucher disease.
  • Orally active and reversible ceramide glucosyltransferase inhibitor
  • Blood-brain barrier permeability
  • Used for research of type I Gaucher disease
  • Restores F508del-CFTR function in cystic fibrosis (CF) bronchial epithelial IB3-1 and CuFi-1 cells
  • Reduces inflammatory response to P. aeruginosa in both CF and non-CF bronchial cells
  • Able to rescue synaptic plasticity deficits, to restore ERKs activation and to counteract hyperexcitability in NPC1-/- mice