Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Miglustat hydrochloride | 210110-90-0 | MFCD00269940 | 98.0% | 255.74 g/mol | C10H22ClNO4 | 10 MG
SDP

Catalog No. 5000294943
Encompass_Preferred

Miglustat hydrochloride is the hydrochloride salt of miglustat, a reversible ceramide glucosyltransferase inhibitor used as a research standard in studies such as type I Gaucher disease. It is provided as an analytical/research reagent with defined purity and storage instructions.

  • Orally active reversible inhibitor of ceramide glucosyltransferase.
  • Intended for research and analytical use, including Gaucher disease studies.
  • Purity 98.0% as supplied.
  • Molecular formula C10H22ClNO4; molecular weight 255.74 g/mol.
  • Store at -20°C under nitrogen; in solvent, store at -80°C up to 6 months.
  • Pack size 10 MG.

Catalog No. 50-002-94943 Supplier Medchemexpress LLC Supplier No. HY17020A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Miglustat hydrochloride is the hydrochloride salt of miglustat, a reversible ceramide glucosyltransferase inhibitor used as a research standard in studies such as type I Gaucher disease. It is provided as an analytical/research reagent with defined purity and storage instructions.

  • Orally active reversible inhibitor of ceramide glucosyltransferase.
  • Intended for research and analytical use, including Gaucher disease studies.
  • Purity 98.0% as supplied.
  • Molecular formula C10H22ClNO4; molecular weight 255.74 g/mol.
  • Store at -20°C under nitrogen; in solvent, store at -80°C up to 6 months.
  • Pack size 10 MG.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.