Learn More
Medchemexpress LLC Migoprotafib | 2377352-49-1 | 98.81% | C25H26N8O | 100 MG
SDP
Catalog No. 5000355893
Encompass_Preferred
Migoprotafib (GDC-1971) is a SHP2 inhibitor that inhibits the MAPK/ERK signaling pathway and exhibits antitumor activity. It is for research use only.
  • Inhibits the MAPK/ERK signaling pathway
  • Exhibits antitumor activity
  • Inhibits proliferation of RIT1 cells
  • Demonstrates antitumor efficacy in RIT1 xenograft mouse models
Catalog No. 50-003-55893 Supplier Medchemexpress LLC Supplier No. HY144903100MG
Migoprotafib (GDC-1971) is a SHP2 inhibitor that inhibits the MAPK/ERK signaling pathway and exhibits antitumor activity. It is for research use only.
  • Inhibits the MAPK/ERK signaling pathway
  • Exhibits antitumor activity
  • Inhibits proliferation of RIT1 cells
  • Demonstrates antitumor efficacy in RIT1 xenograft mouse models