Learn More
Medchemexpress LLC Migoprotafib (GDC-1971) | 2377352-49-1 | 98.8% | 454.53 g·mol⁻1 | C25H26N8O | 5 MG
SDP
Catalog No. 5000221438
Encompass_Preferred
Migoprotafib (GDC-1971) is an orally bioavailable, allosteric inhibitor of the protein tyrosine phosphatase SHP2 that inhibits the MAPK/ERK signaling pathway and has demonstrated antitumor activity in preclinical models. Supplied as a high-purity research reagent for laboratory studies.
  • Allosteric SHP2 inhibition with demonstrated cellular activity.
  • Inhibits MAPK/ERK signaling relevant to tumor growth.
  • Orally bioavailable in in vivo studies, suitable for pharmacology research.
  • High purity suitable for research applications (98.8%).
  • Available in small-mass quantities for compound screening and dosing studies.
Catalog No. 50-002-21438 Supplier Medchemexpress LLC Supplier No. HY1449035MG
Add to Cart
Add to Cart
Migoprotafib (GDC-1971) is an orally bioavailable, allosteric inhibitor of the protein tyrosine phosphatase SHP2 that inhibits the MAPK/ERK signaling pathway and has demonstrated antitumor activity in preclinical models. Supplied as a high-purity research reagent for laboratory studies.
  • Allosteric SHP2 inhibition with demonstrated cellular activity.
  • Inhibits MAPK/ERK signaling relevant to tumor growth.
  • Orally bioavailable in in vivo studies, suitable for pharmacology research.
  • High purity suitable for research applications (98.8%).
  • Available in small-mass quantities for compound screening and dosing studies.